Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,5-Diazabicyclo[4.3.0]Non-5-Ene

Catalog No. 33471500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-471-500ML 500 mL
33-471-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 33-471-500ML Supplier Sigma Aldrich Supplier No. 33471500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) is an amidine base commonly used in the base-mediated eliminations, condensations, esterifications, isomerizations, carboxylations and carbonylations.It is used in the preparation of supertetrahedral chalcogenide clusters and single crystals of polymer-chalcogenide composites.It also acts as a catalyst for the regioselective Friedel-Crafts C-acylation of pyrroles.Amidine base used for dehydrohalogenation reactions to olefins

Specifications

CAS 3001-72-7
Density 1.005 g/mL at 25°C
Boiling Point 95°C to 98°C7.5 mmHg, lit.)
Molecular Formula C7H12N2
Refractive Index n20/D 1.519; n20/D 1.521
Linear Formula C7H12N2
MDL Number MFCD00005554
Quantity 500 mL
Synonym DBN
Molecular Weight (g/mol) 124.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.