Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,5-Diazabicyclo[4.3.0]Non-5-Ene

Catalog No. 1365815G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-658-15G 5 g
13-658-1100G 100 g
13-658-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Danger
Catalog No. 13-658-15G Supplier Sigma Aldrich Supplier No. 1365815G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,5-Diazabicyclo[4.3.0]non-5-ene (DBN) can be used as: A reagent for the synthesis of +a-carbolines from tetrahydro-+a-carbolines via dehydrogenative/decarboxylative aromatization A nucleophilic organocatalyst in the regioselective C-acylation of pyrroles and indoles by Friedel-Crafts acylation reaction.A superbase in the formulation of a ternary liquid-liquid phase changing system, along with hexadecane and hexanol, to capture hydrogen sulfide gas A base for the preparation of 1H-quinazoline-2,4-diones from 2-aminobenzonitriles using supercritical carbon dioxide as a reactant and a solvent. A catalyst for the synthesis of benzothiazolones by the reaction between 2-aminothiophenols and CO2 by cyclocarbonylation reaction via C-S bond formation.

Specifications

CAS 3001-72-7
Density 1.005 g/mL at 25°C
Boiling Point 95°C to 98°C7.5 mmHg, lit.)
Molecular Formula C7H12N2
Refractive Index n20/D 1.519
Linear Formula C7H12N2
MDL Number MFCD00005554
Quantity 5 g
Synonym DBN
Molecular Weight (g/mol) 124.18 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.