Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dibromonaphthalene 98.0+%, TCI America™
SDP

Catalog No. D46605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46605G 5 g
D46601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46605G Supplier TCI America Supplier No. D46605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7351-74-8
Molecular Formula C10H6Br2
Molecular Weight (g/mol) 285.966
MDL Number MFCD00089984
InChI Key CZYAFTZIQWCKOI-UHFFFAOYSA-N
PubChem CID 637960
IUPAC Name 1,5-dibromonaphthalene
SMILES C1=CC2=C(C=CC=C2Br)C(=C1)Br

Specifications

Melting Point 130°C
Color Yellow
Boiling Point 326°C
Quantity 5 g
Formula Weight 285.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dibromonaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.