Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diethynyl-2,4-dimethylbenzene 98.0+%, TCI America™
SDP

Catalog No. D48785G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D48785G 5 g
D48781G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D48785G Supplier TCI America Supplier No. D48785G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1379822-09-9
Molecular Formula C12H10
Molecular Weight (g/mol) 154.212
InChI Key HPFZIHSKAUBEON-UHFFFAOYSA-N
PubChem CID 102363018
IUPAC Name 1,5-diethynyl-2,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1C#C)C#C)C

Specifications

Melting Point 58°C
Quantity 5 g
Formula Weight 154.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Diethynyl-2,4-dimethylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.