Learn More
Medchemexpress LLC 1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate | 125401-63-0 | C41H77NO7 | 100 MG
SDP
Catalog No. 5000375469
Encompass_Preferred
1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate is a lipid utilized in the creation of phospholipid vesicles. It functions alongside other lipids to transform polydisperse vesicles into uniformly sized vesicles through freeze-thaw and extrusion methods. This compound is capable of improving protein encapsulation efficiency for applications in drug delivery and biochemistry.
  • Used in creation of phospholipid vesicles.
  • Transforms polydisperse vesicles into uniformly sized vesicles.
  • Enhances protein encapsulation efficiency.
  • Applicable for drug delivery and biochemical studies.
Catalog No. 50-003-75469 Supplier Medchemexpress LLC Supplier No. HY150220100MG
1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate is a lipid utilized in the creation of phospholipid vesicles. It functions alongside other lipids to transform polydisperse vesicles into uniformly sized vesicles through freeze-thaw and extrusion methods. This compound is capable of improving protein encapsulation efficiency for applications in drug delivery and biochemistry.
  • Used in creation of phospholipid vesicles.
  • Transforms polydisperse vesicles into uniformly sized vesicles.
  • Enhances protein encapsulation efficiency.
  • Applicable for drug delivery and biochemical studies.