Learn More
Medchemexpress LLC 1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate | 125401-63-0 | C41H77NO7 | 500 MG
SDP
Catalog No. 5000374212
Encompass_Preferred
1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate is a lipid utilized in the synthesis of phospholipid vesicles. It works with other lipids to convert polydisperse vesicles into vesicles of uniform size through freeze-thaw and extrusion techniques.
  • Can enhance protein encapsulation efficiency
  • Suitable for drug delivery applications
  • Suitable for biochemical applications
Catalog No. 50-003-74212 Supplier Medchemexpress LLC Supplier No. HY150220500MG
1,5-Dihexadecyl N-(3-carboxy-1-oxopropyl)-L-glutamate is a lipid utilized in the synthesis of phospholipid vesicles. It works with other lipids to convert polydisperse vesicles into vesicles of uniform size through freeze-thaw and extrusion techniques.
  • Can enhance protein encapsulation efficiency
  • Suitable for drug delivery applications
  • Suitable for biochemical applications