Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dihydroxy-4,8-dinitroanthraquinone 96.0+%, TCI America™
SDP

Catalog No. D272725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D272725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D272725G Supplier TCI America Supplier No. D272725G

missing translation for 'validMainframeMsg'

Specifications

CAS 128-91-6
Molecular Formula C14H6N2O8
Quantity 25 g
MDL Number MFCD00035827
InChI Key CUIHODIOWPLCMG-UHFFFAOYSA-N
SMILES OC1=CC=C(C2=C1C(=O)C1=C(C=CC(O)=C1C2=O)[N+]([O-])=O)[N+]([O-])=O
IUPAC Name 1,5-dihydroxy-4,8-dinitro-9,10-dihydroanthracene-9,10-dione
Molecular Weight (g/mol) 330.21
PubChem CID 3090916
Formula Weight 330.21
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Dihydroxy-4,8-dinitroanthraquinone
Show More Show Less
EINECSNumber (4)-0734
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.