Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dihydroxyanthraquinone, tech. 90%, Thermo Scientific™

Catalog No. AAL0084836
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0084836 500 g
AAL0084809 10 g
AAL0084822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0084836 Supplier Thermo Scientific Chemicals Supplier No. L0084836

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-12-4
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001210
InChI Key JPICKYUTICNNNJ-UHFFFAOYSA-N
Synonym anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
PubChem CID 8328
ChEBI CHEBI:37501
IUPAC Name 1,5-dihydroxyanthracene-9,10-dione
SMILES OC1=CC=CC2=C1C(=O)C1=CC=CC(O)=C1C2=O

Specifications

Melting Point ∽280°C (decomposition)
Quantity 500 g
Beilstein 1881718
Merck Index 14,689
Formula Weight 240.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,5-Dihydroxyanthraquinone
EINECSNumber 204-175-6
RTECSNumber CB6630000
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.