Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 98%, Thermo Scientific™

Catalog No. AAH6447503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6447503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6447503 Supplier Thermo Scientific Chemicals Supplier No. H6447503

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5744-59-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00085040
InChI Key PXRXGHUTKHXUGF-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 587757
SMILES CN1N=C(C=C1C)C(O)=O

Specifications

Melting Point 170°C to 174°C
Quantity 1 g
Beilstein 115917
IUPAC Name 1,5-dimethyl-1H-pyrazole-3-carboxylic acid
Formula Weight 140.14
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1,5-Dimethyl-1H-pyrazole-3-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.