Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Dimethyl-2-piperidone 98.0+%, TCI America™
SDP

Catalog No. D27535ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27535ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27535ML Supplier TCI America Supplier No. D27535ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86917-58-0
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD03701098
InChI Key YXHVIDNQBMVYHQ-UHFFFAOYNA-N
PubChem CID 159027
IUPAC Name 1,5-dimethylpiperidin-2-one
SMILES CC1CCC(=O)N(C)C1

Specifications

Color Yellow
Quantity 5 mL
Formula Weight 127.19
Percent Purity ≥98.0% (GC)
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1,5-Dimethyl-2-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.