Learn More
Medchemexpress LLC 1,5-dimethyl-4-(methylamino)-2-phenylpyrazol-3-one | 519-98-2 | MFCD00447796 | 99.8% | C12H15N3O | 10MG
SDP
Catalog No. 5000259746
Encompass_Preferred
4-Methylamino antipyrine is an active metabolite of metamizole with analgesic and antipyretic activity; it inhibits cyclooxygenase (COX) and is supplied for research use as a reference standard in pharmacology, metabolism, and analytical studies.
  • Acts as a cyclooxygenase inhibitor with analgesic and antipyretic effects.
  • Serves as a metabolite reference standard for pharmacokinetic and metabolic studies.
  • High chemical purity (99.8%) suitable for analytical applications.
  • Available in solid and solution formats for experimental flexibility.
  • Small pack sizes convenient for bench-scale research.
Catalog No. 50-002-59746 Supplier Medchemexpress LLC Supplier No. HY13573110MG
4-Methylamino antipyrine is an active metabolite of metamizole with analgesic and antipyretic activity; it inhibits cyclooxygenase (COX) and is supplied for research use as a reference standard in pharmacology, metabolism, and analytical studies.
  • Acts as a cyclooxygenase inhibitor with analgesic and antipyretic effects.
  • Serves as a metabolite reference standard for pharmacokinetic and metabolic studies.
  • High chemical purity (99.8%) suitable for analytical applications.
  • Available in solid and solution formats for experimental flexibility.
  • Small pack sizes convenient for bench-scale research.