Learn More
Northern Safety 1.5 DIOPTER BIFOCAL SFTY GLASS
SDP
Catalog No. NC0992207
Encompass_Preferred
N-Specs� Infusion� Readers 1.5 Diopter Clear Anti-Fog Magnifying Lens Safety Glasses. Get the popular style of the Infusion� with the added features of magnification lenses and anti-fog coating. Soft, cushioned nose piece provides comfortable, non-slip wear. Strategically placed 1.5 magnification lens inserts are out of user's direct line of vision, increasing safety. Anti-fog coating minimizes lens fogging. Impact-resistant polycarbonate lenses provide excellent protection against UV rays. Manufactured in an ISO 9002 Registered facility. Complies with ANSI standards.
This item is not returnable. View return policy
Catalog No. NC0992207 Supplier Northern Safety Supplier No. 28894 15
N-Specs� Infusion� Readers 1.5 Diopter Clear Anti-Fog Magnifying Lens Safety Glasses. Get the popular style of the Infusion� with the added features of magnification lenses and anti-fog coating. Soft, cushioned nose piece provides comfortable, non-slip wear. Strategically placed 1.5 magnification lens inserts are out of user's direct line of vision, increasing safety. Anti-fog coating minimizes lens fogging. Impact-resistant polycarbonate lenses provide excellent protection against UV rays. Manufactured in an ISO 9002 Registered facility. Complies with ANSI standards.