Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Diphenylcarbazide, ≥98% (HPLC), Reag. Ph. Eur., MilliporeSigma™ Supelco™

Catalog No. 111013051 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3051 100 g
11-101-3050 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-3051 Supplier MilliporeSigma Supelco Supplier No. 33152100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,5-Diphenylcarbazide reaction with chromium(VI), has been widely used for the spectro- photometric determination of chromium. It is also an electron donor, and can be used for measuring photosystem 2 activity in subchloroplast fragments.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00003013
Synonym 1,5-Diphenylcarbohydrazide; sym.-Diphenylcarbazide

Specifications

Melting Point 170°C to 175°C; 170°C to 175°C
CAS 140-22-7
Linear Formula C6H5 NHNHCONHNHC6H5
Quantity 100 g
Formula Weight 242.28
Percent Purity ≥98% (HPLC); ≥98.0%
Grade Reag. Ph. Eur.
RTECSNumber FF2750000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.