Learn More
Ace Glass, Inc. 1.5 dram poly-seal vial, cs/2304, sp/144
SDP
Catalog No. 50976408
Encompass
1.5 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention.
Catalog No. 50-976-408 Supplier Ace Glass, Inc. Supplier No. 877915
1.5 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention.