Learn More
MatTek Corporation (Case of 25) No. 1.5, Gridded German glass coverslips, 18 x18 mm
Catalog No. NC1843909
Encompass
No. 1.5, Gridded German glass coverslips, 18 x18 mm
Catalog No. NC1843909 Supplier MatTek Corporation Supplier No. PCS1.51818GRD
No. 1.5, Gridded German glass coverslips, 18 x18 mm