Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Hexadiene-3,4-diol (stabilized with HQ) 95.0+%, TCI America™
SDP

Catalog No. H080710ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H080710ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H080710ML Supplier TCI America Supplier No. H080710ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1069-23-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.14
MDL Number MFCD00081129
InChI Key KUQWZSZYIQGTHT-UHFFFAOYNA-N
Synonym 3,4-Dihydroxy-1,5-hexadiene, Divinylethylene Glycol, 1,2-Divinylglycol
PubChem CID 14042
IUPAC Name hexa-1,5-diene-3,4-diol
SMILES OC(C=C)C(O)C=C

Specifications

Color Yellow
Boiling Point 198°C
Quantity 10 mL
UN Number 2810
Formula Weight 114.14
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,5-Hexadiene-3,4-diol (stabilized with HQ)
RTECSNumber MM2100000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.