Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Hexadiene, 98%

Catalog No. AAA1365036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1365036 500 g
AAA1365014 25 g
AAA1365022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1365036 Supplier Thermo Scientific Chemicals Supplier No. A1365036
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 592-42-7 | C6H10 | 82.146 g/mol

1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

Solubility
Soluble in water(0.17g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-42-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008666
InChI Key PYGSKMBEVAICCR-UHFFFAOYSA-N
Synonym 1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl
PubChem CID 11598
IUPAC Name hexa-1,5-diene
SMILES C=CCCC=C

Specifications

Melting Point -141°C
Density 0.692
Boiling Point 58°C to 60°C
Flash Point −27°C (−16°F)
Odor Turpentine-like
Refractive Index 1.404
Quantity 500 g
UN Number UN2458
Beilstein 1731106
Solubility Information Soluble in water(0.17g/L).
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 1,5-Hexadiene
Show More Show Less
Hazard Category H225-H304-H315-H319-H335
Hazard Statement GHS H Statement
H225-H304-H319-H335
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P331-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEXADIENES
EINECSNumber 209-754-7
RTECSNumber MM1997800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.