Alfa Aesar™ 1,5-Hexadiyne, 50% in pentane

Manufacturer: Alfa Aesar™ 1872004

Quantity 2g

 View more versions of this product

Catalog No. AA1872004

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 628-16-0
    Formula Weight 78.11
    MDL Number MFCD00014924
    UN Number UN3295
    Chemical Name or Material 1,5-Hexadiyne

    Specifications

    Formula Weight 78.11
    Health Hazard 3 GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00014924
    CAS 628-16-0
    Concentration 50% in pentane
    Health Hazard 2 GHS H Statement
    H225-H304-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    May cause drowsiness or dizziness.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
    Quantity 2g
    TSCA TSCA
    Physical Form Liquid
    Health Hazard 1 Danger
    Density 0.805g/mL
    Flash Point −2° (29°F)
    UN Number UN3295
    Storage Note 1
    • Packaged under argon
    Chemical Name or Material 1,5-Hexadiyne

    GHS H Statement
    H225-H304-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P261-P301+P310-P303+P361+P353-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit