Learn More
Cayman Chemical 1 5-IsoquInolIndIol 100mg
SDP
Catalog No. 502973042
Encompass_Preferred
An inhibitor of poly(ADP-ribose) synthetase (PARP1; IC50 = 0.39 μM); can be used with cells in culture and in animals
Catalog No. 50-297-3042 Supplier Cayman Chemical Supplier No. 14438100 MG
An inhibitor of poly(ADP-ribose) synthetase (PARP1; IC50 = 0.39 μM); can be used with cells in culture and in animals