Learn More
Enzo Life Sciences 1,5-Isoquinolinediol (100mg). CAS: 5154-02-9
SDP
Supplier:  Enzo Life Sciences BMLAP1020100
Encompass_Preferred
Potent inhibitor of inducible nitric oxide synthase (iNOS; NOS II) in mouse macrophages. Potent and selective PARP inhibitor. Inhibits nitric oxide (SIN-1) induced PARP activation in rat hepatocytes (at 20µM). Neuroprotectant. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml), dimethyl formamide (25mg/ml) or 100% ethanol (25mg/ml warm). Also soluble in 0.1N NaOH or in methanol; insoluble in water and 0.1N HCl. Long Term Storage: Ambient.
Catalog No. 50-201-0308