Learn More
AdipoGen 1 5-Isoquinolinediol
SDP
Catalog No. 502562391
Encompass_Preferred
Chemical. CAS 5154-02-9. Formula C9H7NO2. MW 161.2. PARP-1 inhibitor. Neuroprotectant. Blocks nitric oxide-induced neuronal toxicity. Potent iNOS inducible nitric oxide synthase/NOS II inhibitor. Stimulates homologous recombination.
Catalog No. 50-256-2391 Supplier AdipoGen Supplier No. AGCR10024M005
Chemical. CAS 5154-02-9. Formula C9H7NO2. MW 161.2. PARP-1 inhibitor. Neuroprotectant. Blocks nitric oxide-induced neuronal toxicity. Potent iNOS inducible nitric oxide synthase/NOS II inhibitor. Stimulates homologous recombination.