Learn More
Medchemexpress LLC 1-(5-isoquinolinesulfonyl)-2-methylpiperazine dihydrochloride | 108930-17-2 | MFCD00036961 | 99.6% | 364.29 g/mol | C14H19Cl2N3O2S | 5 MG
SDP
Catalog No. 5000212020
Encompass_Preferred
Protein kinase inhibitor H-7 dihydrochloride is a small-molecule serine/threonine kinase inhibitor commonly used to inhibit protein kinase C and related kinases in biochemical and cell-based assays. It is supplied as a solid with high reported purity for research applications.
  • Potent protein kinase C (PKC) inhibitor.
  • Broad-spectrum serine/threonine kinase inhibition.
  • Cell-permeable and reversible inhibitor.
  • High reported purity suitable for analytical and biological studies.
  • Stable solid formulation for convenient storage and handling.
  • Recommended storage sealed, away from moisture and light.
Catalog No. 50-002-12020 Supplier Medchemexpress LLC Supplier No. HY1009835MG
Protein kinase inhibitor H-7 dihydrochloride is a small-molecule serine/threonine kinase inhibitor commonly used to inhibit protein kinase C and related kinases in biochemical and cell-based assays. It is supplied as a solid with high reported purity for research applications.
  • Potent protein kinase C (PKC) inhibitor.
  • Broad-spectrum serine/threonine kinase inhibition.
  • Cell-permeable and reversible inhibitor.
  • High reported purity suitable for analytical and biological studies.
  • Stable solid formulation for convenient storage and handling.
  • Recommended storage sealed, away from moisture and light.