Learn More
Medchemexpress LLC 1,5-Methano-1H-3-benzazepine, 2,3,4,5-tetrahydro- | 69718-72-5 | 99.8% | 159.23 | 25 G
SDP
Catalog No. 5000402853
Encompass_Preferred
2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine is a drug intermediate used for the synthesis of various active compounds.
  • Used for research purposes only.
  • Available in multiple quantities from 250 mg to over 100 g.
  • High purity (99.78%).
  • Ships at room temperature within continental US and stores at 4°C, protected from light (or -80°C for 6 months and -20°C for 1 month in solvent, protected from light).
  • Data sheet, COA, SDS, and handling instructions are available.
Catalog No. 50-004-02853 Supplier Medchemexpress LLC Supplier No. 25GHY20007
2,3,4,5-Tetrahydro-1H-1,5-methano-3-benzazepine is a drug intermediate used for the synthesis of various active compounds.
  • Used for research purposes only.
  • Available in multiple quantities from 250 mg to over 100 g.
  • High purity (99.78%).
  • Ships at room temperature within continental US and stores at 4°C, protected from light (or -80°C for 6 months and -20°C for 1 month in solvent, protected from light).
  • Data sheet, COA, SDS, and handling instructions are available.