Learn More
LGC GENOMICS LLC 1.5 ML COLUMN CHUCK
Catalog No. NC2310473
Encompass
Column holder for 1.5 mL syringe style synthesis columns (1 -5 µmol), MerMade 6 and 12 only. Use with column M2-1000, scale dependent on support loading.
Catalog No. NC2310473 Supplier LGC GENOMICS LLC Supplier No. AXM121034
Column holder for 1.5 mL syringe style synthesis columns (1 -5 µmol), MerMade 6 and 12 only. Use with column M2-1000, scale dependent on support loading.