Learn More
USA Scientific Inc 1.5 mL con screw cap tube viol
Catalog No. 502745334
Encompass
1.5 mL conical base microcentrifuge tube, graduated, violet, with integrated TPE cap, 500/pack
Catalog No. 50-274-5334 Supplier USA Scientific Inc Supplier No. 14158305
1.5 mL conical base microcentrifuge tube, graduated, violet, with integrated TPE cap, 500/pack