Learn More
Andwin Scientific 1-5 ML GRADUATED MACRO VOLUME
SDP
Catalog No. 50819439
Encompass_Preferred
The 5 mL pipet tips offer measurement precision with graduations at 0.5 mL increments. This product is compatible with ErgoOne, Gilson Pipetman, or Rainin (not LTS) pipettes. The tips are free of DNA, DNase, and RNase. It comes in 2 bags of 250 tips, totaling to 500 tips.
Catalog No. 50-819-439 Supplier Andwin Scientific Supplier No. 10500000
The 5 mL pipet tips offer measurement precision with graduations at 0.5 mL increments. This product is compatible with ErgoOne, Gilson Pipetman, or Rainin (not LTS) pipettes. The tips are free of DNA, DNase, and RNase. It comes in 2 bags of 250 tips, totaling to 500 tips.