Learn More
Andwin Scientific 1.5 ML GRADUATED MICROCENTRIFU
SDP
Catalog No. 50819705
Encompass
The 1.5 ml copolymer microcentrifuge tube has graduations every 100 l that allow precise sample measurement. This product works for applications involving limited boiling and provides resistance against chemicals. The tube is free of RNase, DNase, DNA, and pyrogen. It has a cap that can be easily opened and a frosted side writing surface that permits easy sample identification. This product comes in a bag of 500 natural colored tubes.
Catalog No. 50-819-705 Supplier Andwin Scientific Supplier No. 14152500
The 1.5 ml copolymer microcentrifuge tube has graduations every 100 l that allow precise sample measurement. This product works for applications involving limited boiling and provides resistance against chemicals. The tube is free of RNase, DNase, DNA, and pyrogen. It has a cap that can be easily opened and a frosted side writing surface that permits easy sample identification. This product comes in a bag of 500 natural colored tubes.