Learn More
Cole-Parmer Argos Technologies Pellet Mixer Microtube, 1.5 mL; 100/Pk
Supplier:  Cole-Parmer 4446821
Encompass_Preferred
Quickly homogenize cells, plant tissue, and DNA pellets within microtube vials. Free of DNAse, RNAse, and pyrogens to eliminate sample contamination
Catalog No. 50-196-0903