Learn More
Andwin Scientific 1.5 ML PP TUBE WITHOUT CAP 10
SDP
Catalog No. 50819699
Encompass_Preferred
The 1.5 ml microcentrifuge tube has an attached flat cap that makes this product ideal for boiling. It also has graduations every 100 l that allow precise sample measurement. The tube is made with polypropylene and is free of RNase, DNase, DNA, and pyrogen. It comes in a bag of 500 tubes in assorted neon colors of blue, green, orange, red, violet, and yellow.
Catalog No. 50-819-699 Supplier Andwin Scientific Supplier No. 14145500
The 1.5 ml microcentrifuge tube has an attached flat cap that makes this product ideal for boiling. It also has graduations every 100 l that allow precise sample measurement. The tube is made with polypropylene and is free of RNase, DNase, DNA, and pyrogen. It comes in a bag of 500 tubes in assorted neon colors of blue, green, orange, red, violet, and yellow.