Learn More
Andwin Scientific 1.5 ML SEAL-RITE MICROCENTRIF
SDP
Supplier:  Andwin Scientific 16155506
Encompass_Preferred
The Seal-Rite 1.5 mL microcentrifuge tube protects samples from evaporation and spillage with its dependable precision sealing system. The tubes will remain tightly closed during the processes of boiling, freezing, centrifugation, and exposure to phenol or chloroform mixture, but can be opened with no difficulty by hand. This product is free of RNase, DNase, DNA, and pyrogens.
Catalog No. 50-819-854