Learn More
CHEMSCIENCE Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 1.5, Individually Serialized, Tolerance ± 0.006 mL
Catalog No. NC2318861
Encompass_Preferred
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 1.5, Individually Serialized, Tolerance ± 0.006 mL
Catalog No. NC2318861 Supplier CHEMSCIENCE Supplier No. 123.302.01B
Pipette Volumetric, With One Mark, Accuracy As Per Class A, Capacity(mL) 1.5, Individually Serialized, Tolerance ± 0.006 mL