Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,5-Naphthalenedisulfonic acid disodium salt hydrate

Catalog No. 250899250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-089-9250G 250 g
25-089-91KG 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 25-089-9250G Supplier Sigma Aldrich Supplier No. 250899250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The liposomal entrapment of 1,5-naphthalenedisulfonic acid disodium salt in phospholipid vesicles has been studied
  • Doubly substituted aromatic dianions of 1,5-naphthalenedisulfonic acid disodium salt has been investigated by collision-induced dissociation, along with infrared multiple photon dissociation/detachment techniques.contains sodium chloride

Specifications

CAS 207569-02-6
Molecular Formula C10H6Na2O6S2 -+ xH2O
Linear Formula C10H6(SO3Na)2 -+ xH2O
Quantity 250 g
MDL Number MFCD00064178
Molecular Weight (g/mol) 332.26 (anhydrous basis)
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.