Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Naphthalenedisulfonic Acid Tetrahydrate 98.0+%, TCI America™
SDP

Catalog No. N076325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N076325G 25 g
N0763500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N076325G Supplier TCI America Supplier No. N076325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 211366-30-2
Molecular Formula C10H16O10S2
Molecular Weight (g/mol) 360.35
MDL Number MFCD00149265
InChI Key ZLBLYGIIADHDKG-UHFFFAOYSA-N
PubChem CID 53249422
IUPAC Name naphthalene-1,5-disulfonic acid tetrahydrate
SMILES O.O.O.O.OS(=O)(=O)C1=CC=CC2=C(C=CC=C12)S(O)(=O)=O

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 288.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Naphthalenedisulfonic Acid Tetrahydrate
EINECSNumber (4)-0467
RTECSNumber QJ5900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.