Learn More
Cayman Chemical 1,5-Anhydro-D-glucitol, Purity: ≥95%, CAS: 154-58-5, 1 g
SDP
Catalog No. 502991803
Encompass_Preferred
A monosaccharide; has been found in a variety of dietary sources including soy, white rice, pasta, vegetables, fish, and tea; renal absorption is blocked by glucose; serum levels are decreased by repeated episodes of hypoglycemia; serum levels have been used as a supplementary glycemic marker to HbA1c in patients with diabetes mellitus
Catalog No. 50-299-1803 Supplier Cayman Chemical Supplier No. 298741 G
A monosaccharide; has been found in a variety of dietary sources including soy, white rice, pasta, vegetables, fish, and tea; renal absorption is blocked by glucose; serum levels are decreased by repeated episodes of hypoglycemia; serum levels have been used as a supplementary glycemic marker to HbA1c in patients with diabetes mellitus