Learn More
Azzota Corp 1.5"", Nickel-Ni, Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 501612968
Encompass_Preferred
1.5", Nickel-Ni, Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 50-161-2968 Supplier Azzota Corp Supplier No. SHC15NI
1.5", Nickel-Ni, Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm