Learn More
Azzota Corp 1.5"" Phosphorus - P Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 501612969
Encompass_Preferred
1.5" Phosphorus - P Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 50-161-2969 Supplier Azzota Corp Supplier No. SHC15P
1.5" Phosphorus - P Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm