Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,5-Pentanediol

Catalog No. P77031KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P77031KG 1 kg
P7703100G 100 g
P7703500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P77031KG Supplier Sigma Aldrich Supplier No. P77031KG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,5-pentanediol can be used:
  • As a monomer for the synthesis of biocompatible polyesters, polycarbonates and polyurethanes
  • As an oxygen precursor for the formation of thin film of zno via atomic layer deposition
  • For the enantioselective convergent synthesis of (+)-spirolaxine methyl ether, αhelicobactericidal agent found in white rot fungiα

Specifications

CAS 111-29-5
Density 0.994 g/mL (at 25°C (literature))
Boiling Point 242°C (lit.)
Molecular Formula C5H12O2
Refractive Index n20/D 1.450 (literature)
Linear Formula HO(CH2)5 OH
MDL Number MFCD00002978
Quantity 1 kg
Synonym Pentamethylene glycol
Molecular Weight (g/mol) 104.15
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.