Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,5-Pentylenediphosphonic Acid 98.0+%, TCI America™
SDP

Catalog No. P23001G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P23001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P23001G Supplier TCI America Supplier No. P23001G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4672-25-7
Molecular Formula C5H14O6P2
Molecular Weight (g/mol) 232.11
MDL Number MFCD00800268
InChI Key QRKINCQKOGXVEO-UHFFFAOYSA-N
Synonym 1,5-Pentanediphosphonic Acid
PubChem CID 2773804
IUPAC Name (5-phosphonopentyl)phosphonic acid
SMILES OP(O)(=O)CCCCCP(O)(O)=O

Specifications

Melting Point 156°C
Color White
Quantity 1 g
UN Number 3261
Formula Weight 232.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,5-Pentylenediphosphonic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.