Learn More
Azzota Corp 1.5" Antimony-Sb, Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Supplier:  Azzota Corp SHC15SB
Encompass_Preferred
1.5" Antimony-Sb, Hollow Cathode Lamp, Self-Reversal, Diameter 37mm, Length 160mm
Catalog No. 50-161-2971