Learn More
Analytical Sales & Services 1.5" Screws for Gen I Reactors using 8x40mm Vials
Catalog No. 5000807049
Encompass
1.5" Screws for Gen I Photoredox and Parallel Synthesis 96 and 24 position reactors (with 9mm spacing)\n\n• Use in place of 1" screws if using 8x40mm vials\n\nQty/Pkg: 100
Catalog No. 50-008-07049 Supplier Analytical Sales & Services Supplier No. 1.5SCREW
Add to Cart
Add to Cart
1.5" Screws for Gen I Photoredox and Parallel Synthesis 96 and 24 position reactors (with 9mm spacing)\n\n• Use in place of 1" screws if using 8x40mm vials\n\nQty/Pkg: 100