Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[5-tert-butyl-2-(4-methylphenyl)pyrazol-3-yl]-3-[5-(pyridin-3-yloxymethyl)-1H-pyrazol-3-yl]urea | 1166393-85-6 | MFCD25977104 | 98.6% | 445.52 g/mol | C24H27N7O2 | 10 MG
SDP

Catalog No. 5000299294
Encompass_Preferred

PF-4618433 is a small-molecule, selective inhibitor of proline-rich tyrosine kinase 2 (PYK2) intended for research use. It exhibits an IC50 of 637 nM against PYK2 and is supplied as a high-purity solid for preclinical assays and exploratory studies in bone-related research.

  • Inhibits PYK2 with a reported IC50 of 637 nM.
  • High purity: 98.6%.
  • Molecular weight 445.52 g/mol.
  • Chemical formula C24H27N7O2.
  • CAS number 1166393-85-6.
  • Supplied as a 10 mg solid for laboratory research.
  • Useful for osteoporosis and bone regeneration studies.

Catalog No. 50-002-99294 Supplier Medchemexpress LLC Supplier No. HY1831210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PF-4618433 is a small-molecule, selective inhibitor of proline-rich tyrosine kinase 2 (PYK2) intended for research use. It exhibits an IC50 of 637 nM against PYK2 and is supplied as a high-purity solid for preclinical assays and exploratory studies in bone-related research.

  • Inhibits PYK2 with a reported IC50 of 637 nM.
  • High purity: 98.6%.
  • Molecular weight 445.52 g/mol.
  • Chemical formula C24H27N7O2.
  • CAS number 1166393-85-6.
  • Supplied as a 10 mg solid for laboratory research.
  • Useful for osteoporosis and bone regeneration studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.