Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[5-(thiophen-2-ylmethoxy)-1H-indol-3-yl]propan-2-amine hydrochloride | 160521-72-2 | MFCD01321066 | 99.8% | 322.85 g/mol | C16H19ClN2OS | 25 MG
SDP

Catalog No. 5000211554
Encompass_Preferred

BW-723C86 is a research-grade small molecule that acts as a selective 5-HT2B receptor agonist, provided as a hydrochloride salt for laboratory applications. It is supplied in purified solid and solution formats for pharmacology and behavioral studies.

  • Selective 5-HT2B receptor agonist for research applications.
  • Purity 99.8%.
  • Hydrochloride salt form for improved handling and solubility.
  • Molecular weight 322.85 g/mol.
  • Chemical formula C16H19ClN2OS.
  • Available as solid and as 10 mM solution in DMSO, including 25 MG pack size.

Catalog No. 50-002-11554 Supplier Medchemexpress LLC Supplier No. HY10136925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BW-723C86 is a research-grade small molecule that acts as a selective 5-HT2B receptor agonist, provided as a hydrochloride salt for laboratory applications. It is supplied in purified solid and solution formats for pharmacology and behavioral studies.

  • Selective 5-HT2B receptor agonist for research applications.
  • Purity 99.8%.
  • Hydrochloride salt form for improved handling and solubility.
  • Molecular weight 322.85 g/mol.
  • Chemical formula C16H19ClN2OS.
  • Available as solid and as 10 mM solution in DMSO, including 25 MG pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.