Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid, Thermo Scientific™

Catalog No. cc73101cb
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC73101CB 250 mg Amber Glass Bottle
CC73101DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC73101CB Supplier Thermo Scientific Chemicals Supplier No. CC73101CB
Add to Cart

Chemical Identifiers

CAS 950603-35-7
Molecular Formula C9H10F3N3O2S
Molecular Weight (g/mol) 281.25
MDL Number MFCD11101440
InChI Key PWQMNLMMMCREOV-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl-1,3,4-thiadiazol-2-yl piperidine-4-carboxylic acid
PubChem CID 33589561
IUPAC Name 1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]piperidine-4-carboxylic acid
SMILES OC(=O)C1CCN(CC1)C1=NN=C(S1)C(F)(F)F

Specifications

Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 281.25
Percent Purity 97%
Chemical Name or Material 1-[5-(trifluoromethyl)-1, 3, 4-thiadiazol-2-yl]piperidine-4-carboxylic acid
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.