Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(5-Trifluoromethyl-pyridin-2-yl)-piperazine, 97%, Thermo Scientific™

Catalog No. SEW04033EB
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
SEW04033EB 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SEW04033EB Supplier Thermo Scientific Chemicals Supplier No. SEW04033EB

Specifications

CAS 132834-58-3
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C10H14F3N3
Quantity 25g
MDL Number MFCD00114708
Synonym 1-5-trifluoromethyl pyridin-2-yl piperazine, 1-5-trifluoromethyl pyrid-2-yl piperazine, 1-5-trifluoromethyl-pyridin-2-yl-piperazine, 1-5-trifluoromethyl-2-pyridinyl piperazine, 1-5-trifluoromethyl-2-pyridinyl-piperazine, 1-5-trifluoromethyl-2-pyridyl piperazine, 1-5-trifluoromethyl pyridine-2-yl piperazine, piperazine, 1-5-trifluoromethyl-2-pyridinyl, 5-trifluoromethyl-2-pyridyl piperazine
InChI Key BNMSJUIMZULLAS-UHFFFAOYSA-P
SMILES FC(F)(F)C1=CC=C([NH+]=C1)N1CC[NH2+]CC1
IUPAC Name 4-[5-(trifluoromethyl)pyridin-1-ium-2-yl]piperazin-1-ium
Molecular Weight (g/mol) 233.24
PubChem CID 667618
Formula Weight 231.22
Percent Purity 97%
Chemical Name or Material 1-(5-Trifluoromethyl-pyridin-2-yl)-piperazine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.