Learn More
Ibidi USA µ-Slide Spheroid Perfusion, ibiTreat
SDP
Catalog No. NC2082306
Encompass
A perfusable channel µ-Slide with 3x7 wells for long-term spheroid cultivation and high-end microscopy. Ideal for the long-term cultivation and perfusion of 3D spheroids. Allowing for flexible preparation for perfusion, spheroids can be transferred or created directly in the wells. Imaging Chamber with excellent optical quality for high-resolution microscopy. Pcs./Box: 15 (individually packed)Surface Modification: ibiTreat: #1.5 polymer coverslip, surface passivation with ibiTreat, sterilized
This item is not returnable. View return policy
Catalog No. NC2082306 Supplier Ibidi USA Supplier No. 80356
A perfusable channel µ-Slide with 3x7 wells for long-term spheroid cultivation and high-end microscopy. Ideal for the long-term cultivation and perfusion of 3D spheroids. Allowing for flexible preparation for perfusion, spheroids can be transferred or created directly in the wells. Imaging Chamber with excellent optical quality for high-resolution microscopy. Pcs./Box: 15 (individually packed)Surface Modification: ibiTreat: #1.5 polymer coverslip, surface passivation with ibiTreat, sterilized