Learn More
MSC Energizer. 390-389TZ Size 390/389, Silver Oxide, Button & Coin Cell Battery 1.55 Volts, Button Tab Terminal, SR1130W, SR54, IEC Regulated
Catalog No. NC9082625
Encompass
Energizer. 390-389TZ Size 390/389, Silver Oxide, Button & Coin Cell Battery 1.55 Volts, Button Tab Terminal, SR1130W, SR54, IEC Regulated
Catalog No. NC9082625 Supplier MSC Supplier No. 76581057
Energizer. 390-389TZ Size 390/389, Silver Oxide, Button & Coin Cell Battery 1.55 Volts, Button Tab Terminal, SR1130W, SR54, IEC Regulated