Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uline 1.5"" X 1.5"" 2ML RECLOS BAG CS

Catalog No. NC1236154
Encompass_Preferred

Uline Inc 1.5" X 1.5" 2ml resealable bag. 1000/carton

Catalog No. NC1236154 Supplier Uline Supplier No. S10819
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Uline Inc 1.5" X 1.5" 2ml resealable bag. 1000/carton

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.