Learn More
Thermco Products Inc -1/51C THERMO PARTIAL IMM
Catalog No. NC1122274
Encompass_Preferred
Thermco’s Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In-Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case. 
Catalog No. NC1122274 Supplier Thermco Products Inc Supplier No. ACC10033
Thermco’s Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In-Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case. 