Learn More
Thermco Products Inc NIST compliant, factory certified glass Spirit thermometer, -1 deg./51 deg.C, 0.2C div, 76mm imm, certified at 0 deg.C, 10 deg.C, 20 deg.C, 30 deg.C, 37 deg.C, 40 deg.C, 50 deg.C, 105 deg.C, 400mm
Catalog No. NC0347706
Encompass_Preferred
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.
Catalog No. NC0347706 Supplier Thermco Products Inc Supplier No. ACC10033BLSFC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.