Learn More
Analytical West Santoprene 3-Stop Tubing, Yellow-Blue-Yellow, (1.52mm), 12/pk
SDP
Catalog No. NC1708535
Encompass
Three-Stop Santoprene tubing has overall length of 400mm (15.75-inches) with 72mm (2.8-inches) between the stops.
Catalog No. NC1708535 Supplier Analytical West Supplier No. PT3220PH2
Three-Stop Santoprene tubing has overall length of 400mm (15.75-inches) with 72mm (2.8-inches) between the stops.